siRNA Details
Gene Symbol: WTIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080436.1 | NP_001073905.1 | 5 | 1019 | ||
XM_006723014.3 | XP_006723077.1 | 5 | 1052 | |||
XM_011526452.2 | XP_011524754.1 | 5 | 1059 | |||
XM_011526453.2 | XP_011524755.1 | 5 | 453 | |||
GenBank mRNA | AK130059.1 | - | 4 | 357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
