siRNA Details
Gene Symbol: ZNF473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026574.1 | XP_016882063.1 | 3 | 3468 | ||
GenBank mRNA | AB032967.1 | - | 5 | 2225 | ||
AF454744.1 | - | 4 | 2180 | |||
AK054671.1 | - | 1 | 1367 | |||
AK292452.1 | - | 5 | 2118 | |||
AK302293.1 | - | 4 | 2075 | |||
AL080143.1 | - | 1 | 533 | |||
BC018612.1 | - | 5 | 2148 | 4634056 | ||
BX648093.1 | - | 5 | 2269 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
