siRNA Details
Gene Symbol: ZNF473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026574.1 | XP_016882063.1 | 3 | 4311 | ||
GenBank mRNA | AB032967.1 | - | 5 | 3068 | ||
AF454744.1 | - | 4 | 3023 | |||
AL080143.1 | - | 1 | 1376 | |||
BX648093.1 | - | 5 | 3112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
