siRNA Details
Gene Symbol: FAM71E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308429.1 | NP_001295358.1 | 5 | 1199 | ||
NM_138411.2 | NP_612420.1 | 5 | 1151 | |||
XM_011526405.2 | XP_011524707.2 | 5 | 849 | |||
XM_011526406.2 | XP_011524708.2 | 5 | 801 | |||
GenBank mRNA | AK125745.1 | - | 5 | 772 | ||
AK131032.1 | - | 5 | 658 | |||
BC004941.1 | - | 5 | 610 | |||
BC012203.1 | - | 5 | 752 | |||
BC071737.1 | - | 5 | 1151 | 5192796 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
