siRNA Details
Gene Symbol: CFAP74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304360.1 | NP_001291289.1 | 18 | 2185 | ||
XM_011542333.2 | XP_011540635.2 | 18 | 2102 | |||
XM_011542335.1 | XP_011540637.1 | 13 | 1614 | |||
XM_011542336.2 | XP_011540638.1 | 13 | 3349 | |||
XM_017002641.1 | XP_016858130.1 | 19 | 2311 | |||
XM_017002642.1 | XP_016858131.1 | 19 | 2311 | |||
XM_017002643.1 | XP_016858132.1 | 19 | 2311 | |||
GenBank mRNA | AB051538.1 | - | 15 | 1857 | ||
BC139754.1 | - | 17 | 2062 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
