siRNA Details
Gene Symbol: CFAP74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304360.1 | NP_001291289.1 | 15, 16 | 1924 | ||
XM_011542333.2 | XP_011540635.2 | 15, 16 | 1841 | |||
XM_011542335.1 | XP_011540637.1 | 10, 11 | 1353 | |||
XM_011542336.2 | XP_011540638.1 | 10, 11 | 3088 | |||
XM_017002641.1 | XP_016858130.1 | 16, 17 | 2050 | |||
XM_017002642.1 | XP_016858131.1 | 16, 17 | 2050 | |||
XM_017002643.1 | XP_016858132.1 | 16, 17 | 2050 | |||
GenBank mRNA | AB051538.1 | - | 12, 13 | 1596 | ||
BC139754.1 | - | 14, 15 | 1801 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
