siRNA Details
Gene Symbol: DMRTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032110.2 | NP_115486.1 | 2 | 810 | ||
XM_011541937.2 | XP_011540239.1 | 1 | 1599 | |||
GenBank mRNA | AJ301580.1 | - | 1 | 304 | ||
BC136282.1 | - | 1 | 566 | 9020269 | ||
BC143800.1 | - | 1 | 566 | 9052314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
