siRNA Details
Gene Symbol: DMRTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032110.2 | NP_115486.1 | 3 | 1850 | ||
XM_011541937.2 | XP_011540239.1 | 2 | 2639 | |||
GenBank mRNA | BC136282.1 | - | 2 | 1594 | 9020269 | |
BC143800.1 | - | 2 | 1600 | 9052314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
