siRNA Details
Gene Symbol: SYPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040709.1 | NP_001035799.1 | 3 | 604 | ||
XM_011541283.2 | XP_011539585.1 | 3 | 604 | |||
XM_011541284.2 | XP_011539586.1 | 3 | 604 | |||
XM_011541285.1 | XP_011539587.1 | 3 | 604 | |||
GenBank mRNA | AB158472.1 | - | 3 | 238 | ||
AI983207.1 | - | 3 | 457 | |||
AK131459.1 | - | 3 | 604 | |||
AK289513.1 | - | 3 | 538 | |||
AK293082.1 | - | 3 | 480 | |||
AK302566.1 | - | 3 | 524 | |||
DB291596.1 | - | 3 | 524 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
