siRNA Details
Gene Symbol: SYPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040709.1 | NP_001035799.1 | 4 | 763 | ||
XM_011541283.2 | XP_011539585.1 | 4 | 763 | |||
XM_011541284.2 | XP_011539586.1 | 4 | 763 | |||
XM_011541285.1 | XP_011539587.1 | 4 | 763 | |||
GenBank mRNA | AB158472.1 | - | 4 | 397 | ||
AK131459.1 | - | 4 | 763 | |||
AK289513.1 | - | 4 | 697 | |||
AK293082.1 | - | 4 | 639 | |||
AK301509.1 | - | 1 | 21 | |||
AK302566.1 | - | 4 | 683 | |||
BC113102.1 | - | 1 | 172 | 40077145 | ||
BC113103.1 | - | 1 | 160 | 40077149 | ||
DN831287.1 | - | 1 | 155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
