siRNA Details
Gene Symbol: ZNF697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080470.1 | NP_001073939.1 | 3 | 1624 | ||
XM_005271315.3 | XP_005271372.1 | 3 | 1650 | |||
XM_011542416.2 | XP_011540718.1 | 3 | 4629 | |||
XM_017002783.1 | XP_016858272.1 | 3 | 1885 | |||
GenBank mRNA | AK311787.1 | - | 1 | 343 | ||
BC007260.1 | - | 1 | 565 | |||
BC033126.1 | - | 1 | 565 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
