siRNA Details
Gene Symbol: FLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002016.1 | NP_002007.1 | 2 | 637 | ||
GenBank mRNA | M60495.1 | - | 1 | 475 | ||
M60499.1 | - | 1 | 36 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
