siRNA Details
Gene Symbol: PPFIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304331.1 | NP_001291260.1 | 30 | 6019 | ||
NM_001304332.1 | NP_001291261.1 | 29 | 5992 | |||
XM_006711586.2 | XP_006711649.1 | 30 | 5910 | |||
XM_011510070.2 | XP_011508372.1 | 18 | 3939 | |||
XM_017002606.1 | XP_016858095.1 | 29 | 5883 | |||
XM_017002607.1 | XP_016858096.1 | 29 | 5775 | |||
XM_017002609.1 | XP_016858098.1 | 29 | 5914 | |||
GenBank mRNA | AB020704.1 | - | 12 | 3148 | ||
AK023365.1 | - | 11 | 2129 | |||
AK124535.1 | - | 5 | 4089 | |||
AK126185.1 | - | 1 | 3255 | |||
AK126624.1 | - | 17 | 4587 | |||
EF428334.1 | - | 35 | 5886 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
