siRNA Details
Gene Symbol: PPFIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304331.1 | NP_001291260.1 | 27 | 3683 | ||
NM_001304332.1 | NP_001291261.1 | 26 | 3656 | |||
XM_006711586.2 | XP_006711649.1 | 27 | 3713 | |||
XM_011510070.2 | XP_011508372.1 | 15 | 1742 | |||
XM_017002606.1 | XP_016858095.1 | 26 | 3686 | |||
XM_017002607.1 | XP_016858096.1 | 26 | 3578 | |||
XM_017002608.1 | XP_016858097.1 | 26 | 3686 | |||
XM_017002609.1 | XP_016858098.1 | 26 | 3578 | |||
GenBank mRNA | AB020704.1 | - | 9 | 812 | ||
AF034801.1 | - | 11 | 1207 | |||
AK023365.1 | - | 8 | 853 | |||
AK124535.1 | - | 3 | 1170 | |||
AK126624.1 | - | 14 | 2251 | |||
AK295757.1 | - | 21 | 2411 | |||
BC132923.1 | - | 14 | 2193 | 40146945 | ||
BC132925.1 | - | 14 | 2193 | 40146947 | ||
BC144262.1 | - | 13 | 2166 | 9052782 | ||
EF428334.1 | - | 32 | 3550 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
