siRNA Details
Gene Symbol: THSD7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316349.1 | NP_001303278.1 | 3 | 991 | ||
GenBank mRNA | AB051466.1 | - | 1 | 615 | ||
AK299929.1 | - | 4 | 1127 | |||
BC019344.1 | - | 1 | 72 | |||
BC033125.1 | - | 1 | 72 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
