siRNA Details
Gene Symbol: THSD7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316349.1 | NP_001303278.1 | 11 | 2503 | ||
XM_017005049.1 | XP_016860538.1 | 5 | 980 | |||
GenBank mRNA | AB051466.1 | - | 9 | 2127 | ||
BC019344.1 | - | 9 | 1584 | |||
BC033125.1 | - | 9 | 1584 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
