siRNA Details
Gene Symbol: NYAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020864.1 | NP_065915.1 | 2 | 318 | ||
XM_005246708.3 | XP_005246765.1 | 3 | 1189 | |||
XM_011511524.1 | XP_011509826.1 | 3 | 1189 | |||
XM_017004564.1 | XP_016860053.1 | 2 | 742 | |||
GenBank mRNA | AB040919.1 | - | 2 | 226 | ||
BC131727.1 | - | 2 | 248 | |||
DA230522.1 | - | 2 | 557 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
