siRNA Details
Gene Symbol: MARCH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020814.2 | NP_065865.1 | 4 | 3526 | ||
GenBank mRNA | AL353945.1 | - | 1 | 148 | ||
BC048793.1 | - | 4 | 1600 | |||
BC098448.1 | - | 4 | 1955 | 6209260 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
