siRNA Details
Gene Symbol: MARCH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020814.2 | NP_065865.1 | 4 | 4333 | ||
GenBank mRNA | AB037820.1 | - | 4 | 4337 | ||
AL353945.1 | - | 1 | 957 | |||
BC048793.1 | - | 4 | 2405 | |||
BC098448.1 | - | 4 | 2768 | 6209260 | ||
U79281.1 | - | 1 | 660 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
