siRNA Details
Gene Symbol: LOC100996693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286811.1 | NP_001273740.1 | 2 | 223 | ||
XM_011510469.2 | XP_011508771.1 | 3 | 2704 | |||
GenBank mRNA | DA086253.1 | - | 1 | 29 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
