siRNA Details
Gene Symbol: GRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080423.3 | NP_001073892.3 | 24 | 4580 | ||
XM_011534141.2 | XP_011532443.1 | 24 | 4662 | |||
XM_011534142.2 | XP_011532444.1 | 24 | 4574 | |||
XM_011534143.2 | XP_011532445.1 | 23 | 4536 | |||
XM_011534145.2 | XP_011532447.1 | 18 | 4308 | |||
XM_011534146.2 | XP_011532448.1 | 23 | 5141 | |||
GenBank mRNA | AB051506.2 | - | 24 | 4566 | ||
AF052177.1 | - | 1 | 189 | |||
AK024507.1 | - | 11 | 3202 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
