siRNA Details
Gene Symbol: FAM198A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129908.2 | NP_001123380.2 | 2 | 1105 | ||
GenBank mRNA | AK091001.1 | - | 2 | 1035 | ||
AK311059.1 | - | 2 | 778 | |||
AL117530.1 | - | 1 | 348 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
