siRNA Details
Gene Symbol: MYH15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014981.1 | NP_055796.1 | 17 | 1910 | ||
XM_011512559.2 | XP_011510861.1 | 18 | 2031 | |||
XM_017005922.1 | XP_016861411.1 | 8 | 1175 | |||
GenBank mRNA | AB023217.3 | - | 17 | 1910 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
