siRNA Details
Gene Symbol: CCDC39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181426.1 | NP_852091.1 | 1, 2 | 207 | ||
GenBank mRNA | AK304270.1 | - | 1, 2 | 186 | ||
AL122120.1 | - | 7, 8 | 964 | |||
DC399927.1 | - | 1, 2 | 207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
