siRNA Details
Gene Symbol: CCDC39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181426.1 | NP_852091.1 | 3 | 402 | ||
GenBank mRNA | AK304270.1 | - | 3 | 381 | ||
AL122120.1 | - | 9 | 1159 | |||
DC399927.1 | - | 3 | 402 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
