siRNA Details
Gene Symbol: STXBP5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308330.1 | NP_001295259.1 | 3 | 414 | ||
NM_014980.2 | NP_055795.1 | 3 | 414 | |||
XM_006713825.3 | XP_006713888.1 | 3 | 584 | |||
XM_011513328.2 | XP_011511630.1 | 2 | 261 | |||
XM_011513331.2 | XP_011511633.1 | 3 | 411 | |||
XM_011513333.2 | XP_011511635.1 | 3 | 411 | |||
XM_017007530.1 | XP_016863019.1 | 3 | 345 | |||
XM_017007531.1 | XP_016863020.1 | 3 | 399 | |||
XM_017007532.1 | XP_016863021.1 | 3 | 348 | |||
XM_017007534.1 | XP_016863023.1 | 3 | 411 | |||
XM_017007535.1 | XP_016863024.1 | 3 | 602 | |||
GenBank mRNA | AB023223.1 | - | 3 | 381 | ||
AK296544.1 | - | 3 | 412 | |||
BC022029.1 | - | 3 | 392 | |||
DC324331.1 | - | 3 | 414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
