siRNA Details
Gene Symbol: VWA5B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320373.1 | NP_001307302.1 | 17 | 3512 | ||
NM_138345.2 | NP_612354.1 | 19 | 4100 | |||
XM_011513288.2 | XP_011511590.1 | 20 | 4739 | |||
XM_011513289.2 | XP_011511591.1 | 20 | 4822 | |||
XM_011513290.2 | XP_011511592.1 | 20 | 4285 | |||
XM_011513291.2 | XP_011511593.1 | 19 | 4948 | |||
XM_011513292.2 | XP_011511594.1 | 19 | 4939 | |||
GenBank mRNA | AL834499.1 | - | 13 | 2932 | ||
BC001200.1 | - | 14 | 3279 |

More Information
Other Species Targeted

