siRNA Details
Gene Symbol: VWA5B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320373.1 | NP_001307302.1 | 13 | 1959 | ||
NM_138345.2 | NP_612354.1 | 15 | 2547 | |||
XM_011513288.2 | XP_011511590.1 | 16 | 3186 | |||
XM_011513289.2 | XP_011511591.1 | 16 | 3269 | |||
XM_011513290.2 | XP_011511592.1 | 16 | 2732 | |||
XM_011513291.2 | XP_011511593.1 | 15 | 3395 | |||
XM_011513292.2 | XP_011511594.1 | 15 | 3386 | |||
GenBank mRNA | AK293885.1 | - | 13 | 1959 | ||
AL834499.1 | - | 9 | 1379 | |||
BC001200.1 | - | 10 | 1726 | |||
BC136609.1 | - | 15 | 2547 | 9020599 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
