siRNA Details
Gene Symbol: CISD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008388.4 | NP_001008389.1 | 3 | 1459 | ||
GenBank mRNA | BC032300.1 | - | 3 | 1399 | 5105935 | |
BX537971.1 | - | 3 | 1397 | |||
DB565768.1 | - | 1 | 266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
