siRNA Details
Gene Symbol: USP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019050.2 | NP_061923.2 | 16 | 3255 | ||
XM_005263073.3 | XP_005263130.1 | 15 | 3129 | |||
XM_005263075.2 | XP_005263132.1 | 15 | 3144 | |||
XM_005263076.2 | XP_005263133.1 | 15 | 3141 | |||
XM_005263077.3 | XP_005263134.1 | 16 | 3294 | |||
XM_006714240.1 | XP_006714303.1 | 14 | 2330 | |||
XM_011532037.2 | XP_011530339.1 | 16 | 3307 | |||
XM_011532038.2 | XP_011530340.1 | 15 | 2584 | |||
XM_017008312.1 | XP_016863801.1 | 17 | 3421 | |||
XM_017008313.1 | XP_016863802.1 | 16 | 3106 | |||
XM_017008314.1 | XP_016863803.1 | 16 | 3105 | |||
XM_017008315.1 | XP_016863804.1 | 16 | 3175 | |||
XM_017008316.1 | XP_016863805.1 | 14 | 2993 | |||
XM_017008317.1 | XP_016863806.1 | 15 | 3118 | |||
XM_017008318.1 | XP_016863807.1 | 16 | 3240 | |||
GenBank mRNA | AB037771.1 | - | 9 | 1704 | ||
AJ583824.1 | - | 13 | 2186 | |||
BX648339.1 | - | 14 | 2343 | |||
CR749490.1 | - | 17 | 3371 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
