siRNA Details
Gene Symbol: USP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019050.2 | NP_061923.2 | 18 | 3476 | ||
XM_005263073.3 | XP_005263130.1 | 17 | 3350 | |||
XM_005263075.2 | XP_005263132.1 | 17 | 3365 | |||
XM_005263076.2 | XP_005263133.1 | 17 | 3362 | |||
XM_005263077.3 | XP_005263134.1 | 17 | 3418 | |||
XM_006714240.1 | XP_006714303.1 | 16 | 2551 | |||
XM_011532037.2 | XP_011530339.1 | 18 | 3528 | |||
XM_011532038.2 | XP_011530340.1 | 17 | 2805 | |||
XM_017008312.1 | XP_016863801.1 | 19 | 3642 | |||
XM_017008313.1 | XP_016863802.1 | 18 | 3327 | |||
XM_017008314.1 | XP_016863803.1 | 18 | 3326 | |||
XM_017008315.1 | XP_016863804.1 | 18 | 3396 | |||
XM_017008316.1 | XP_016863805.1 | 16 | 3214 | |||
XM_017008317.1 | XP_016863806.1 | 16 | 3242 | |||
XM_017008318.1 | XP_016863807.1 | 17 | 3364 | |||
GenBank mRNA | AB037771.1 | - | 11 | 1925 | ||
AJ583824.1 | - | 15 | 2407 | |||
AK022799.1 | - | 2 | 143 | |||
BX648339.1 | - | 16 | 2564 | |||
CR749490.1 | - | 19 | 3592 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
