siRNA Details
Gene Symbol: MAST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164664.1 | NP_001158136.1 | 10 | 1535 | ||
NM_001290226.1 | NP_001277155.1 | 8 | 987 | |||
NM_001290227.1 | NP_001277156.1 | 8 | 759 | |||
NM_001297651.1 | NP_001284580.1 | 7 | 1019 | |||
NM_015183.2 | NP_055998.1 | 9 | 728 | |||
XM_006714606.3 | XP_006714669.1 | 8 | 981 | |||
XM_006714610.2 | XP_006714673.1 | 7 | 968 | |||
XM_011543382.2 | XP_011541684.1 | 12 | 1627 | |||
XM_011543384.2 | XP_011541686.1 | 7 | 2686 | |||
XM_011543385.2 | XP_011541687.1 | 7 | 3806 | |||
XM_011543386.2 | XP_011541688.1 | 7 | 637 | |||
XM_017009447.1 | XP_016864936.1 | 13 | 1708 | |||
XM_017009448.1 | XP_016864937.1 | 11 | 1591 | |||
XM_017009449.1 | XP_016864938.1 | 8 | 1026 | |||
XM_017009450.1 | XP_016864939.1 | 8 | 720 | |||
XM_017009451.1 | XP_016864940.1 | 8 | 720 | |||
XM_017009452.1 | XP_016864941.1 | 8 | 4035 | |||
XM_017009453.1 | XP_016864942.1 | 7 | 10850 | |||
GenBank mRNA | AK057601.1 | - | 7 | 972 | ||
AK131421.1 | - | 9 | 1069 | |||
AK296707.1 | - | 8 | 724 | |||
AK299538.1 | - | 8 | 869 | |||
AK299706.1 | - | 7 | 1018 | |||
AK309559.1 | - | 7 | 916 | |||
AY830839.1 | - | 9 | 729 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
