siRNA Details
Gene Symbol: MAST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164664.1 | NP_001158136.1 | 16 | 2295 | ||
NM_001290226.1 | NP_001277155.1 | 14 | 1747 | |||
NM_001290227.1 | NP_001277156.1 | 14 | 1519 | |||
NM_001297651.1 | NP_001284580.1 | 13 | 1779 | |||
NM_015183.2 | NP_055998.1 | 15 | 1488 | |||
XM_006714606.3 | XP_006714669.1 | 14 | 1741 | |||
XM_006714610.2 | XP_006714673.1 | 13 | 1728 | |||
XM_011543382.2 | XP_011541684.1 | 18 | 2387 | |||
XM_011543384.2 | XP_011541686.1 | 13 | 3446 | |||
XM_011543385.2 | XP_011541687.1 | 13 | 4566 | |||
XM_011543386.2 | XP_011541688.1 | 13 | 1397 | |||
XM_017009447.1 | XP_016864936.1 | 19 | 2468 | |||
XM_017009448.1 | XP_016864937.1 | 17 | 2351 | |||
XM_017009449.1 | XP_016864938.1 | 14 | 1786 | |||
XM_017009450.1 | XP_016864939.1 | 14 | 1480 | |||
XM_017009451.1 | XP_016864940.1 | 14 | 1480 | |||
XM_017009452.1 | XP_016864941.1 | 14 | 4795 | |||
XM_017009453.1 | XP_016864942.1 | 13 | 11610 | |||
GenBank mRNA | AB002301.1 | - | 5 | 531 | ||
AK057601.1 | - | 13 | 1732 | |||
AK296707.1 | - | 14 | 1484 | |||
AK299538.1 | - | 14 | 1629 | |||
AK299706.1 | - | 13 | 1778 | |||
AY830839.1 | - | 15 | 1489 | |||
BX648796.1 | - | 1 | 40 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
