siRNA Details
Gene Symbol: ANKRD31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164443.1 | NP_001157915.1 | 7 | 941 | ||
XM_011543298.2 | XP_011541600.1 | 7 | 963 | |||
XM_011543299.1 | XP_011541601.1 | 7 | 941 | |||
XM_011543300.2 | XP_011541602.1 | 7 | 963 | |||
XM_011543301.2 | XP_011541603.1 | 7 | 963 | |||
XM_011543302.2 | XP_011541604.1 | 5 | 499 | |||
XM_017009317.1 | XP_016864806.1 | 7 | 858 | |||
XM_017009318.1 | XP_016864807.1 | 7 | 963 | |||
XM_017009320.1 | XP_016864809.1 | 4 | 484 | |||
XM_017009322.1 | XP_016864811.1 | 7 | 963 | |||
GenBank mRNA | AK097510.1 | - | 7 | 941 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
