siRNA Details
Gene Symbol: N4BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015111.1 | NP_055926.1 | 5 | 1783 | ||
XM_006714834.3 | XP_006714897.1 | 5 | 1813 | |||
XM_011534473.1 | XP_011532775.1 | 5 | 1832 | |||
XM_011534474.1 | XP_011532776.1 | 5 | 1832 | |||
GenBank mRNA | AB002339.1 | - | 4 | 1544 | ||
AK127224.1 | - | 1 | 807 | |||
AK295663.1 | - | 2 | 651 | |||
BC053323.1 | - | 5 | 1783 | 6585266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
