siRNA Details
Gene Symbol: MYLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012418.3 | NP_001012418.2 | 13 | 2367 | ||
XM_005249078.3 | XP_005249135.2 | 13 | 2332 | |||
XM_005249079.2 | XP_005249136.1 | 13 | 2225 | |||
XM_006715082.3 | XP_006715145.1 | 12 | 3662 | |||
XM_011514567.2 | XP_011512869.1 | 13 | 2455 | |||
XM_017010819.1 | XP_016866308.1 | 13 | 2264 | |||
GenBank mRNA | AK122581.1 | - | 5 | 1371 | ||
AK127360.1 | - | 5 | 1268 | |||
BX647582.1 | - | 13 | 2366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
