siRNA Details
Gene Symbol: PCDHB18P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_001281.2 | 1 | 622 | |||
GenBank mRNA | AF152528.1 | - | LIT2076 PCDH-psi2 |
1 | 404 | |
AF217743.1 | - | 1 | 241 | |||
AK095496.1 | - | 1 | 591 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
