siRNA Details
Gene Symbol: LRRTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015564.2 | NP_056379.1 | 2 | 1113 | ||
GenBank mRNA | AB007876.1 | - | 2 | 1065 | ||
AK291064.1 | - | 2 | 1084 | |||
AK297332.1 | - | 2 | 659 | |||
AY182026.1 | - | 1 | 656 | |||
BC126408.1 | - | 2 | 684 | 8992124 | ||
BC126410.1 | - | 2 | 684 | 8992126 | ||
BC143926.1 | - | 2 | 684 | 9052444 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
