siRNA Details
Gene Symbol: LRRTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015564.2 | NP_056379.1 | 2 | 1306 | ||
GenBank mRNA | AK291064.1 | - | 2 | 1277 | ||
AK297332.1 | - | 2 | 852 | |||
BC126408.1 | - | 2 | 877 | 8992124 | ||
BC126410.1 | - | 2 | 877 | 8992126 | ||
BC143926.1 | - | 2 | 877 | 9052444 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
