siRNA Details
Gene Symbol: PPP1R18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134870.1 | NP_001128342.1 | 4 | 2815 | ||
NM_133471.3 | NP_597728.1 | 3 | 4461 | |||
GenBank mRNA | AB073600.1 | - | 4 | 2706 | ||
AB075829.1 | - | 3 | 3879 | |||
AK097089.1 | - | 3 | 2614 | |||
AK124880.1 | - | 3 | 4461 | |||
AK125639.1 | - | 3 | 2623 | |||
AK126182.1 | - | 2004 | ||||
AL832318.1 | - | 3 | 1742 | |||
AL832742.1 | - | 3 | 3210 | |||
BC006176.1 | - | 3 | 1269 | |||
BC066644.1 | - | 3 | 2773 | 5575643 | ||
BX647622.1 | - | 4 | 2715 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
