siRNA Details
Gene Symbol: PPP1R18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134870.1 | NP_001128342.1 | 2, 3 | 1844 | ||
NM_133471.3 | NP_597728.1 | 1, 2 | 3490 | |||
GenBank mRNA | AB073600.1 | - | 2, 3 | 1735 | ||
AB075829.1 | - | 1, 2 | 2908 | |||
AK097089.1 | - | 1, 2 | 1643 | |||
AK124880.1 | - | 1, 2 | 3490 | |||
AK126182.1 | - | 811 | ||||
AL832318.1 | - | 1, 2 | 771 | |||
AL832742.1 | - | 1, 2 | 2241 | |||
BC006176.1 | - | 1, 2 | 298 | |||
BC066644.1 | - | 1, 2 | 1802 | 5575643 | ||
BX647622.1 | - | 2, 3 | 1744 | |||
DQ988947.1 | - | 1, 2 | 285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
