siRNA Details
Gene Symbol: PPP1R18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134870.1 | NP_001128342.1 | 4 | 2237 | ||
NM_133471.3 | NP_597728.1 | 3 | 3883 | |||
GenBank mRNA | AB073600.1 | - | 4 | 2128 | ||
AB075829.1 | - | 3 | 3301 | |||
AK097089.1 | - | 3 | 2036 | |||
AK124880.1 | - | 3 | 3883 | |||
AK125639.1 | - | 3 | 2045 | |||
AK126182.1 | - | 1426 | ||||
AL832318.1 | - | 3 | 1164 | |||
AL832742.1 | - | 3 | 2633 | |||
BC006176.1 | - | 3 | 691 | |||
BC066644.1 | - | 3 | 2195 | 5575643 | ||
BX647622.1 | - | 4 | 2137 | |||
DQ988947.1 | - | 3 | 678 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
