siRNA Details
Gene Symbol: ANO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001891.3 | NP_001001891.2 | 22 | 2613 | ||
XM_011511263.2 | XP_011509565.1 | 22 | 2561 | |||
XM_017004230.1 | XP_016859719.1 | 21 | 2542 | |||
GenBank mRNA | AK057322.1 | - | 2 | 124 | ||
AY617079.1 | - | 22 | 2614 | |||
BC028162.1 | - | 2 | 123 | |||
BC038852.1 | - | 2 | 115 | |||
BC047903.1 | - | 2 | 140 | |||
CD672346.1 | - | 3 | 447 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
