siRNA Details
Gene Symbol: CLVS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010852.3 | NP_001010852.2 | 5 | 2133 | ||
XM_017010266.1 | XP_016865755.1 | 5 | 3835 | |||
GenBank mRNA | AK095527.1 | - | 5 | 2137 | ||
AK295158.1 | - | 5 | 595 | |||
BC101376.1 | - | 5 | 1667 | 40025886 | ||
BC101377.2 | - | 5 | 1669 | 40025889 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
