siRNA Details
Gene Symbol: ABRACL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021243.2 | NP_067066.1 | 3 | 281 | ||
XM_017011151.1 | XP_016866640.1 | 3 | 266 | |||
GenBank mRNA | AF116682.1 | - | 3 | 216 | ||
AF161398.1 | - | 3 | 262 | |||
BC014953.1 | - | 3 | 154 | |||
BG563686.1 | - | 3 | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
