siRNA Details
Gene Symbol: VWDE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135924.1 | NP_001129396.1 | 12 | 2932 | ||
XM_005249641.1 | XP_005249698.1 | 9 | 2046 | |||
XM_011515174.1 | XP_011513476.1 | 12 | 2609 | |||
XM_011515175.1 | XP_011513477.1 | 12 | 2609 | |||
XM_017011828.1 | XP_016867317.1 | 10 | 2264 | |||
XM_017011829.1 | XP_016867318.1 | 9 | 1933 | |||
XM_017011830.1 | XP_016867319.1 | 12 | 2609 | |||
GenBank mRNA | AK027618.1 | - | 1 | 519 | ||
BC144316.1 | - | 12 | 2647 | |||
BC144319.1 | - | 9 | 2084 | 9052839 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
