siRNA Details
Gene Symbol: LINC00525 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038407.1 | 3 | 572 | |||
GenBank mRNA | BC007354.2 | - | 3 | 572 | 3616854 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
