siRNA Details
Gene Symbol: LOC541473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003602.3 | 5 | 816 | |||
GenBank mRNA | BC022013.2 | - | 5 | 770 | ||
BC067783.1 | - | 5 | 789 |
Gene Symbol: LOC100101148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104007.1 | 4 | 678 | |||
GenBank mRNA | BC070189.1 | - | 4 | 678 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
