siRNA Details
Gene Symbol: NOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138400.1 | NP_612409.1 | 9 | 2234 | ||
GenBank mRNA | AY576779.1 | - | 9 | 2234 | ||
BC007902.2 | - | 5 | 525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
