siRNA Details
Gene Symbol: RAB19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008749.2 | NP_001008749.2 | 3 | 524 | ||
XM_005249996.3 | XP_005250053.1 | 3 | 807 | |||
XM_005249997.3 | XP_005250054.1 | 3 | 790 | |||
XM_005249998.3 | XP_005250055.1 | 3 | 785 | |||
XM_017012207.1 | XP_016867696.1 | 3 | 720 | |||
GenBank mRNA | BC140796.1 | - | 2 | 394 | 9021664 | |
DN996121.1 | - | 3 | 498 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
